Learn More
Cayman Chemical CL2SN38dIchLRoacetIc acI 5mg
SDP
Supplier:  Cayman Chemical 282155 MG
Encompass_Preferred
A reactive drug-linker molecule containing SN-38 and a maleimidocaproyl linker; cytotoxic to LoVo human colon adenocarcinoma cells (IC50 = 5.3 nM) when linked to hMN-14 as an ADC; increases survival in a colon carcinoma mouse model of lung metastasis when linked to hMN-14
Catalog No. 50-293-9695