Learn More
AdipoGen Cladospirone bisepoxide
SDP
Supplier:  AdipoGen BVT0065M001
Encompass_Preferred
Chemical. CAS 152607-03-9 and 155866-40-3. Formula C20H14O7. MW 366.3. Isolated from Sphaeropsidales sp. Antibiotic. Antibacterial and antifungal. Antitumor compound. Germination inhibitor.
Catalog No. 50-256-2125