Learn More
Selleck Chemical LLC Cladribine 25mg 4291-63-8 2-CdA, 2-chlorodeoxyadenosine
SDP
Supplier:  Selleck Chemical LLC S119925MG
Encompass_Preferred
Cladribine (2-CdA, 2-chlorodeoxyadenosine) is an adenosine deaminase inhibitor for U266, RPMI8226, and MM1.S cells with IC50 of approximately 2.43 ?M, 0.75 ?M, and 0.18 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-689-7