Learn More
Selleck Chemical LLC Cladribine S1199-1g
SDP
Supplier:  Selleck Chemical LLC S11991G
Encompass_Preferred
Cladribine is an adenosine deaminase inhibitor for U266 RPMI8226 and MM1 S cells with IC50 of approximately 2 43 M 0 75 M and 0 18 M respectively
Catalog No. 50-311-1065