Learn More
Enterprise Technology Solutions Claire Manufacturing Co Cleaner,f/StainlessSteel,Cleans/Polishes/Protects,Fingerprint&SmudgeResist,Oil-Based,15oz,12/CT,Gray
SDP
Supplier:  Enterprise Technology Solutions CGCSW841
Encompass_Preferred
Sprayway Oil-Based Stainless Steel Cleaner is specially formulated to effectively clean stainless steel fixtures, equipment or trim. It cleans, polishes and protects surfaces without scouring. Easily remove old or heavy accumulation of greasy film, food spatter or oil stains. Cleaner resists fingerprints, grease, water spatter and smudges, and it doesn't leave behind streaks. Simply spray and wipe clean with a microfiber cloth (not included).
Catalog No. 50-008-32287