Learn More
Chemglass Life Sciences Chemglass Clamp, 19/38 Standard Taper Joint, Blue PTFE
SDP
Supplier:  Chemglass Life Sciences CG145P19
Encompass_Preferred
The Chemglass Life Sciences CG-145-P-19 is a color-coded PTFE clamp designed for 19/38 standard taper glass joints. It secures standard taper connections and provides chemical inertness from its PTFE construction.
  • PTFE material with steel core.
  • Color: Blue.
  • Fits 19/38 standard taper joints.
  • Chemically resistant and inert.
  • Withstands temperatures up to 250°C.
  • Sold individually (each).
Catalog No. NC3735043