Learn More
Wilmad Labglass CLAMP EXT SGL ADJ MED 2 1/4IN
Supplier:  Wilmad Labglass LG4637102
Encompass
Clamp, Extension, Three Prong, Dual Adjustment. Maximum Grip Size: 2- 1/4 in. (57mm); Maximum Load: 15kg; Overall Length: 8- 3/4 in. (22cm), Clamp Extension: 5- 1/8 in. (13cm)
Catalog No. 50-801-133