Learn More
World Precision Instrument Clamp for MM1 and MM1-3
SDP
Supplier:  World Precision Instrument MM1C
Encompass
  • Mount electrodes on miniature manipulators
  • Opens 0-4mm
Catalog No. 50-252-1077