Learn More
Wilmad Labglass CLAMP KETTLE 500ML1L
Supplier:  Wilmad Labglass LG7317100
Encompass
Clamp, Reaction Kettle. For Flat Flange. Fits Flange Size: 4- 1/2 in. Clamp is cast bronze ring w/ black anodized finish. Three spring-activated clamping arms fit beneath the flange of the flask/reaction vessel body.
Catalog No. 50-801-086