Learn More
Wilmad Labglass CLAMP MCCARTER 80MM
Supplier:  Wilmad Labglass LG7316104
Encompass
Clamp, McCarter. Clamp Size: 80mm. Stainless steel horseshoe-shaped clamp used to connect flat flanges together.
Catalog No. 50-802-065