Learn More
Wilmad Labglass CLAMP PINCH LOCKING SZ 28
Supplier:  Wilmad Labglass LG1045110
Encompass
Clamp, Pinch, Spherical Joints, Stainless Steel. Clamp Size: 28. This clamp is for use w/ spherical joints and is manufactured from stainless steel. Supplied w/ a screw type locking devise.
Catalog No. 50-802-201