Learn More
Wilmad Labglass CLAMP QUICK RELEASE 100MM
Supplier:  Wilmad Labglass LG4632102
Encompass
Clamp, Quick Release, For Duran Flange. Fits Flange ID: 100mm. Quick release clamp made of stainless steel. Has screw adjustment for tension control to flange.
Catalog No. 50-801-950