Learn More
Wilmad Labglass CLAMP ST METAL SZ 29
Supplier:  Wilmad Labglass LG1006106
Encompass
Joint Clamp, Metal, for Standard Taper joints, Fits Joint Sizes: 29/26, 29/42. Clamp is lacquer coated to avoid scratching glass. Joints are secure under moderate pressure to 200degreeC.
Catalog No. 50-802-258