Learn More
Chemglass Life Sciences CLAMP THREE PRONG EXTENSION
SDP
Supplier:  Chemglass Life Sciences SAN4958302MSR/QMS51078
Encompass_Preferred
Clamp, Three Prong Extension, Medium, Dual Adjust, 1/2" OD x 9" Arm Length
Catalog No. NC3930564