Learn More
Chemglass Life Sciences Clamp, Three-Prong Extension, Open Yoke, Large, Single Adjust, 10.24in OAL
SDP
Supplier:  Chemglass Life Sciences CG921003
Encompass_Preferred
Clamp, Three-Prong Extension, Open Yoke, Large, Single Adjust, 10 1/2in OAL. Nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation, with an open yoke to allow full use of the clamp depth. Three-prong clamps are available in single and dual adjustment. The small clamp has a 1¿ (25mm) maximum grip size with a 1/4¿ (6mm) O.D. extension arm. The medium clamp has a 1 1/2¿ (38mm) maximum grip size with a 1/2¿ (13mm) O.D. extension arm. The large clamp has a 2 1/2¿ (64mm) maximum grip size with a 5/8¿ (16mm) O.D. extension arm. Complete with removable vinyl and fiberglass covers for easy cleaning.
Catalog No. CG921003