Learn More
Grainger Clamp Universal Platform Flasks Tube
Supplier:  Grainger 14N132
Encompass_Preferred
The H1000-MR is a horizontal laminar flow clean bench made by Benchmark Scientific. It provides HEPA-filtered, particle-free air to protect samples from contamination. Key features include a stainless steel interior, LED lighting, and adjustable airflow speed. It has overall dimensions of 48"W x 24"D x 35"H. This clean bench is designed for applications like PCR setup, tissue culture, and more. It meets ISO 5 cleanroom standards.
Catalog No. 50-275-0276