Learn More
Med Vet International Lorna-Edna Towel Forceps, 3.5 in., Non-Perforating Teeth, Von Klaus German Surgical Steel
Supplier:  Med Vet International VK1175709
Encompass_Preferred
Lorna Towel Forceps have a non perforating clamp that is used to secure instruments and surgical material. Lorna Towel Clamps can also be known as a Edna Towel Clamp. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1719