Learn More
Lkt Laboratories Clarithromycin, ≥95%, 100MG
SDP
Supplier:  Lkt Laboratories C4502100MG
Encompass_Preferred
Macrolide; protein translation inhibitor. CAS: 81103-11-9. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC9492493