Learn More
Medchemexpress LLC Clarithromycin-d3 | 959119-22-3 | 99.90% | 750.97 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17508S15MG
Encompass_Preferred
Clarithromycin-d3 is the deuterium labeled Clarithromycin. It has a broad spectrum of antimicrobial activity and inhibits CYP3A4-catalyzed triazolam alpha-hydroxylation with an IC50 (Ki) value of 56 (43) μM. It also significantly inhibits the HERG potassium current and affects autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86569