Learn More
AdipoGen Clarithromycin N-oxide
SDP
Supplier:  AdipoGen CDXC0310M010
Encompass_Preferred
Chemical. CAS 118074-07-0. Formula C38H69NO14. MW 763.95. Synthetic Biologically incactive urinary metabolite of Clarithromycin oxidized by ozone, a macrolide antibiotic. Compound can be used as analytical reference material.
Catalog No. 50-256-1390