Learn More
MSC Vermont Gage Minus Pin Gage, Tool Steel, 0.1550 in. Diameter, 2 in. Long, 1 ea.
Supplier: MSC 88815501
This Class X Minus Pin Gage is a precision instrument designed for the calibration of measuring equipment or for direct gaging of parts. Constructed from high-quality tool steel, this gage features a nominal diameter of 0.1550 in. and a total length of 2 in.
- Class X precision tolerance of -0.00004 in.
- Tool steel construction provides durability and stability
- NIST traceable for reliable measurement accuracy
- Features a 4 micro finish for superior surface quality
- Roundness within 1/2 of tolerance
- Certificate of accuracy included with each unit