Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 0.948 in. Dia., 1 ea.
Supplier:  MSC 88994801
Encompass
This Class ZZ plug gage is designed for precise measurements and inspection tasks, such as verifying hole sizes, calculating distances between holes, gaging slot widths, and checking locations. It is also suitable for setting micrometers and other measuring devices. This gage is constructed from high-quality 52100 tool steel and is heat-treated to 60 to 62 Rc for enhanced durability and wear resistance.
  • Class ZZ tolerance of +0.0002 in.
  • Constructed from 52100 tool steel
  • Heat-treated to 60 to 62 Rc hardness
  • Centerless lapped with a 10 micro-finish or better
  • Includes a certificate of accuracy with NIST traceability
  • Electro-etched with size and direction of tolerance
  • Smooth ground ends
Catalog No. 50-005-15941