Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, 0.0290 in. Diameter, Steel, 1 ea.
Supplier:  MSC 89002901
Encompass
This Class ZZ steel plug and pin gage is a precision inspection tool designed for measuring hole sizes. It features a nominal diameter of 0.0290 in. and an overall length of 2 in., providing reliable accuracy for verifying tolerances in various shop applications.
  • Tolerance of +0.0000/-0.0002 in.
  • Hardness rating of 60/65 Rc
  • Surface finish of 10 microinch or better
  • NIST traceable with included certificate of accuracy
  • Meets ISO 17025:2005 standards
  • Suitable for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-15765