Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, 0.8190 in. Diameter, Tool Steel, 2 in. OAL, 1 ea.
Supplier: MSC 89081905
This Class ZZ plug and pin gage is designed for precision inspection and measurement applications. Constructed from durable tool steel, this reversible gage features a 0.8190 in. diameter and a 2 in. overall length, providing reliable performance for setting micrometers, verniers, and other measurement devices.
- Tolerance of 0.0002 in. with a minus tolerance direction
- Hardened to 60/65 Rc
- Finish of 10 microinch or better
- Roundness within 0.0001 in. (0.0025 mm)
- Includes certificate of accuracy with NIST traceability
- Size and direction of tolerance marked on the gage