Learn More
Lakeside Manufacturing Inc Classic Anesthesia Cart, Blue with Telegraph Gray, (1) Push Button Lock, (6) Drawers
Supplier:  Lakeside Manufacturing Inc C630P2B
Encompass_Preferred
Lakeside Classic Anesthesia carts allow for storage, organization, and security of all required supplies. The Lakeside C-630-P-2B Anesthesia Cart is blue with telegraph gray, has a push button lock, and (6) drawers. This cart provides secure, mobile storage for medical supplies. Its steel construction is durable with an epoxy finish. This Lakeside cart's 6-drawers with push button lock offers an extra layer of security, while its outrigger stabilizing frame with full size poly extruded corner bumpers protects walls and furniture. This cart has twin ergonomic vertical push handles and an easy-to-clean and sanitize molded top tray. Equipped with quiet non-marking swivel casters - 2 ea. with brake, 1 ea. with directional lock. Modify or custom-build with no minimum orders. Superior lead times. Made in the USA.
Catalog No. 50-288-4317