Learn More
Lakeside Manufacturing Inc Classic Emer. Cart 6-Drw Brkwy
Supplier:  Lakeside Manufacturing Inc C630B1R
Encompass_Preferred
Provide easy access to equipment, medications and supplies on the fly. The Lakeside C-630-B-1R Classic Emergency Cart with 6-drawers and breakaway lock has a solid red finish. This cart has a drawer configuration of 4-3" (76), 1-6" (152), 1-12" (305). Its twin poly handles make for comfort and easy of maneuverability. This emergency cart's plastic top prevents supplies from falling off cart, while its premium 5" (127) swivel casters (2 locking, 1 directional) provide smooth maneuverability around tight spaces. Its full extension ball-bearing drawer glides for silent and smooth operation. This cart's breakaway seal and lock bar ensure supplies are sterilized. Its durable dual-wall steel construction and antimicrobial powder coating for robust usage. This medical cart's solid plastic corner bumpers protect walls and furniture. Available in 23 standard colors. Made in the USA.
Catalog No. 50-288-4351