Learn More
Cayman Chemical ClastoLactacystIn lacton 100UG
SDP
Supplier:  Cayman Chemical 70988100 UG
Encompass_Preferred
Active metabolite of lactacystin, a widely used selective inhibitor of the 20S proteasome, with at least 10 times better activity; irreversibly alkylates subunit X of the 20S proteasome
Catalog No. 50-289-5452