Learn More
Medchemexpress LLC Clathrin-IN-4 | 762242-59-1 | C17H18Br2N2O | 100 MG

Supplier: Medchemexpress LLC HY152218100MG
Clathrin-IN-4 is an analogue of Wiskostatin that acts as a potent inhibitor of clathrin-mediated endocytosis with an IC50 of 2.1 μM. It also functions as a dynamin I GTPase activity inhibitor with an IC50 of 9.1 μM.
- Purity of 99.54%
- Appears as a solid, white to off-white
- Ships at room temperature in continental US
- Recommended storage at 4°C, protected from light
- Soluble in DMSO at 90 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.