Learn More
OriGene Claudin 1 (CLDN1) CytoSection, 25 Slides
SDP
Supplier:  OriGene TS404466P5
Encompass_Preferred
Transient overexpression of CLDN1 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3627494