Learn More
Genetex Inc Claudin 3 Monoclonal Antibody [HMV309] HistoMAX™ (GTX639933) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639933
Encompass_Preferred
The Claudin 3 antibody [HMV309] HistoMAX™ (GTX639933) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human fallopian tube mucosa tissue using GTX639933 Claudin 3 antibody [HMV309] HistoMAX. Strong membranous claudin 3 staining of epithelial cells.
Catalog No. NC3669305