Learn More
ABclonal Technology Claudin 5 Rabbit mAb
SDP
Supplier:  ABclonal Technology A25830100UL
Encompass_Preferred
This gene encodes a member of the claudin family Claudins are integral membrane proteins and components of tight junction strands Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets Mutations in this gene have been found in patients with velocardiofacial syndrome Alternative splicing results in multiple transcript variants encoding distinct isoforms
Catalog No. 50-000-09144