Learn More
Cell Signaling Technology Claudin-6 (E7U2O) XP(R) Rabbit mAb (BSA and Azide Free) 100 ug
SDP
Supplier:  Cell Signaling Technology 59648SF
Encompass
Claudin-6 (E7U2O) XP(R) Rabbit mAb (BSA and Azide Free) 100 ug
Catalog No. 50-273-2700