Learn More
OriGene Claudin 9 (CLDN9) CytoSection
SDP
Supplier:  OriGene TS408701P5
Encompass_Preferred
Transient overexpression of CLDN9 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3881578