Learn More
Medchemexpress LLC Clavulanate (lithium) | 61177-44-4 | 99.9% | C8H8LiNO5 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYA0256B10MM/1ML
Encompass_Preferred
Clavulanate lithium is a potent β-lactamase inhibitor that acts as an antibiotic. While it has weak antibacterial activity alone, in combination with beta-lactam antibiotics, it prevents antibiotic inactivation by microbial lactamase. This product is for research use only.
  • Potent β-lactamase inhibitor
  • Acts as an antibiotic
  • Prevents antibiotic inactivation by microbial lactamase
  • White to off-white solid
  • For research use only
Catalog No. 50-004-32469