Learn More
Selleck Chemical LLC Clavulanic acid E4472-5MG
SDP
Supplier:  Selleck Chemical LLC E44725MG
Encompass_Preferred
Clavulanic acid is a naturally occurring powerful bacterial -lactamase inhibitor The beta-lactam ring found in clavulanic acid binds to the active site of beta-lactamase resulting in enzyme inactivation and enhancing the antibacterial activity of beta-lactam antibiotics like amoxicillin
Catalog No. 50-000-39656