Learn More
Cayman Chemical ClavulnatepotasIum salt 10mg
SDP
Supplier:  Cayman Chemical 1945610 MG
Encompass_Preferred
A β-lactamase inhibitor that is effective against Ambler class A β-lactamases (IC50 values range from 12 to 60 nM); commonly used with other antibiotics that would be inactivated by β-lactamases
Catalog No. 50-298-3545