Learn More
Cayman Chemical ClazamycIn AhydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 402241 MG
Encompass_Preferred
An antibiotic; active against a variety of bacteria, including S. aureus, B. anthracis, B. subtilis, E. coli, and P. aeruginosa (MICs = 100, 6.25, 100, 50, and 25 UG/ml, respectively)
Catalog No. 50-294-2153