Learn More
Aviva Systems Biology Corp CLCA2 Antibody - C-terminal region (ARP35195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLCA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CLCA2. Alternate names: CACC, CACC3, CLCRG2, CaCC-3, FLJ97885ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-451