Learn More
Aviva Systems Biology Corp CLDN15 Antibody - middle region (ARP33636_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CLDN15. Alternate names: FLJ42715, MGC19536ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-55104