Learn More
ABclonal Technology CLDN17 Rabbit pAb
SDP
Supplier:  ABclonal Technology A151511000UL
Encompass_Preferred
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. This gene is intronless and is clustered with CLDN8 on chromosome 21q22.11.
Catalog No. 50-001-07432