Learn More
Trilink Bio Technologies Inc CleanCap FLuc mRNA, 100 ug
SDP
Supplier:  Trilink Bio Technologies Inc L7602100
Encompass
The FLuc mRNA will express a luciferase protein, originally isolated from the firefly, Photinus pyralis. FLuc is commonly used in mammalian cell culture to measure both gene expression and cell viability. It emits bioluminescence in the presence of the substrate, luciferin.
Catalog No. NC1360329