Learn More
Essendant Simple Green Extreme Aircraft and Precision Equipment Cleaner, Neutral Scent, 55 gal Drum
Supplier:  Essendant SMP13455
Encompass_Preferred
Extreme Simple Green Aircraft and Precision Cleaner was specifically designed with long term metal cleaning in mind It s a non-flammable water-based cleaner with an added corrosion inhibitor that provides an extra layer of protection when cleaning delicate metals rubbers and plastics Its combined high soil capacity superior grease-cutting ability and clean-rinsing formula prevents redeposition - ensuring fast effective and high precision cleaning This formula can be used on aluminum carbon fiber high tech alloys structural metals paints protective coatings and more Extreme avoids oxidizing finishes rusting surfaces and degrading plastics making it ideal for maintaining structural metals hoses seals and factory finishes Application- Degreaser/Cleaner Applicable Material- Enamel Fiberglass Metal Plastic Rubber Chemical Compound- Water based non-ionic surfactant Dirt Types- Grease Grime Lubricants Oil-Based Soil
Catalog No. 50-000-26498