Learn More
Essendant Simple Green Pro HD Heavy-Duty Cleaner, Unscented, 1 gal Bottle, 4/Carton
Supplier:  Essendant SMP13421
Encompass_Preferred
Simple Green Pro HD is a heavy-duty cleaner and degreaser that breaks down tough grease and grime This powerful formula removes dirt and stains from a wide variety of surfaces including engines car exteriors tools machinery floors walls and more Formulated with a corrosion inhibitor making it ideal for metals It s also non-degrading to plastics and clean rinsing The cost-effective concentrate can be custom diluted for specific cleaning jobs and used in pressure washers carpet cleaners floor scrubbers ultrasonics dip tanks and other cleaning equipment Pro HD provides an effective alternative to solvents and other harsh cleaning chemicals without sacrificing performance Application- Degreaser/Cleaner Applicable Material- Ceramic Laminate Metal Plastic Rubber Chemical Compound- Triethanolamine Ethoxylated Alcohol Propylene Glycol Butyl Ether Tetrapotassium Pyrophosphate Potassium Silicate Dirt Types- Dirt Dust Grease Oil
Catalog No. 50-000-28705