Learn More
Essendant Virex II 256 One-Step Disinfectant Cleaner Deodorant Mint, 1 gal, 4 Bottles/CT
Supplier:  Essendant DVO04332
Encompass_Preferred
A one-step, quaternary-based disinfectant cleaner and deodorant for use in facilities where cleaning and prevention of cross-contamination are critical. Neutral, no-rinse formula will not dull, pit or soften floor finishes. Bactericidal, virucidal and fungicidal. Meets hospital standards for decontaminating blood and body fluids.
Catalog No. NC1077854