Learn More
Essendant Rust-Oleum Professional Pressure Washer Concentrate Multi-Purpose, 1 gal Jug, 4/Carton
Supplier:  Essendant RST352251
Encompass_Preferred
Rust-Oleum Multi-Purpose Pressure Washer Concentrate is the Pro s choice to make outdoor cleaning jobs easier The fast-acting bleach-free formula is the ideal solution for effectively removing grease grime stains from mold mildew and algae oil fungus and more from a variety of exterior surfaces no scrubbing required Rust-Oleum Multi-Purpose Pressure Washer Concentrate contains anti-corrosive agents to safeguard pressure washer parts Application- Outdoor Surface Cleaner Applicable Material- Brick Cement Block Concrete Driveways Gutters Masonry Roofing Siding Chemical Compound- Proprietary Blend of Detergents Foaming Surfactants Dye Physical Form- Liquid
Catalog No. 50-000-27370