Learn More
Essendant Rust-Oleum Professional Cleaner Degreaser Concentrated, 1 gal Bottle, 4/Carton
Supplier:  Essendant RST352261
Encompass_Preferred
Rust-Oleum Concentrated Cleaner Degreaser is the ideal solution for industrial cleaning jobs Powerful non-toxic formula tackles grease grime oil and more with maximum cleaning performance Multi-purpose cleaner and degreaser is ideal for various industrial cleaning tasks including equipment machinery engine parts tools and more Fabric safe and leaves no residue behind Application- Combustion Engine Parts Construction Equipment Industrial Equipment Machinery Tools Applicable Material- Canvas Fabric/Textiles Fiberglass Glass/Ceramics Metal Plastic Sealed Asphalt/Blacktop Sealed Cement/Concrete Sealed Masonry Wood Chemical Compound- Fatty Alcohol Ethoxylate Potassium Hydroxide Sodium Metasilicate Dirt Types- Adhesives/Glues Grease Inorganic Soils Latex Paint Oil Organic Soils Oxidation Soot Tar Wax
Catalog No. 50-000-27456