Learn More
Essendant Rust-Oleum General Purpose Cleaner Degreaser, 5 gal Bucket
Supplier:  Essendant RST352257
Encompass_Preferred
Rust-Oleum PRO Concentrated Cleaner Degreaser is a heavy duty formula for industrial cleaning applications It is designed to quickly dissolve grease and grime on most non-porous surfaces For use on cleaning tools machinery engine parts construction equipment fiberglass unfinished and finished concrete masonry asphalt and more This concentrated commercial strength formula safely and easily removes many stains and substances such as food and drink stains dried latex paint paint overspray tape residue glue and adhesive acid rain bird droppings smoke damage tree sap grease and oil marker and crayon pet stains brake dust fireplace soot oxidation lipstick scuff marks blood stains sun tan oil tar and wax chewing gum soap scum mildew stains shoe polish and more An all-purpose formula (1 5 cups of concentrate to 1 gallon of water) cleans and degreases appliances mini-blinds canvas chrome silver brass copper porcelain plastic fiberglass linoleum alumin
Catalog No. 50-000-29119