Learn More
Essendant CLEANER8OZA-STAT SPRY
Supplier:  Essendant END097000
Encompass_Preferred
Specially formulated pump spray safely cleans all types of electronic equipment Picks up dust and gently cleans away fingerprints Stays where you spray it so it won t run into sensitive areas Once applied it repels dust and helps prevent buildup of static charges Oil-free wax-free and ammonia-free Contains alcohol Application- Electronics/Antistatic Cleaner Applicable Material- Ceramics Fiberglass Glass Laminates Metal Plastic Chemical Compound- Ethanol Isopropyl Alcohol Dirt Types- Debris Dust Grease Fingerprints Soil Smudges
Catalog No. 50-000-35689