Learn More
Essendant CLEANERAPC32OZ6CS
Supplier:  Essendant EOPPL97066CT
Encompass_Preferred
ECOS PRO Orange Plus is an all-purpose cleaner and degreaser that effectively lifts dirt grime scum grease and adhesive residue with the power of citrus plants Use it to clean and fight odors on water-safe surfaces This formula is made using sustainable plant-powered ingredients Made without dyes gray water and septic safe Features a refreshing citrus scent Not tested on animals Manufactured in a facility that is LEED Zero Carbon LEED Zero Waste and LEED Zero Energy Certified Application- Multi Purpose Cleaner/Degreaser Applicable Material- Water-Safe Hard Surfaces Chemical Compound- Water Caprylyl/Myristyl Glucoside Potassium Sorbate Citric Acid Alcohol Denat Limonene Dirt Types- Adhesive Residue Dirt Grease Grime Scum
Catalog No. 50-000-35319