Learn More
Essendant CLEANERFOAMDETERGENT
Supplier:  Essendant DVO57772100
Encompass_Preferred
EnduroSafe cleaner is a soft metal-safe heavy duty chlorinated alkaline product designed for extended contact cleaning in a wide range of food and beverage processing operations Application- Degreaser/Cleaner Applicable Material- Soft Metals Chemical Compound- Sodium Tripolyphosphate N N-Dimethyltetradecylamine N-Oxide Dirt Types- Fat Deposits General Soils in Food Plants Oily Soils
Catalog No. 50-000-21809